Assay Details
| Gene Symbol: AS3MT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001113391.4 | XP_001113391.3 | 2406 | 93 | ||
| GenBank mRNA | JU326434.1 | - | 444 | 93 | ||
| JU326435.1 | - | 444 | 93 | |||
| JV066571.1 | - | 464 | 93 | |||
| JV066572.1 | - | 464 | 93 | |||
More Information
Set Membership: |
|

AS3MT
Genomic Map