Assay Details
| Gene Symbol: GORASP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131326.2 | XP_014986812.1 | 876 | 85 | ||
| XM_015131327.2 | XP_014986813.1 | 876 | 85 | |||
| XM_015131328.2 | XP_014986814.1 | 1056 | 85 | |||
| XM_015131329.2 | XP_014986815.1 | 580 | 85 | |||
| GenBank mRNA | JU323180.1 | - | 860 | 85 | ||
| JU473484.1 | - | 861 | 85 | |||
| JV045558.1 | - | 856 | 85 | |||
| JV554815.1 | - | 867 | 85 | |||
More Information
Set Membership: |
|

GORASP1
Genomic Map