Assay Details
| Gene Symbol: MOB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257738.1 | NP_001244667.1 | 566 | 122 | ||
| XM_015131736.2 | XP_014987222.1 | 524 | 122 | |||
| XM_015131737.2 | XP_014987223.1 | 674 | 122 | |||
| XM_015131738.2 | XP_014987224.1 | 497 | 122 | |||
| GenBank mRNA | JU333859.1 | - | 566 | 122 | ||
| JV046675.1 | - | 575 | 122 | |||
More Information
Set Membership: |
|

MOB4
Genomic Map