Assay Details
| Gene Symbol: IL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044727.1 | NP_001038192.1 | 172 | 98 | ||
| GenBank mRNA | DQ890030.1 | - | 165 | 98 | ||
| DQ890031.1 | - | 165 | 98 | |||
| DQ890032.1 | - | 165 | 98 | |||
| DQ890033.1 | - | 165 | 98 | |||
| DQ890034.1 | - | 165 | 98 | |||
| DQ890035.1 | - | 165 | 98 | |||
| DQ890036.1 | - | 165 | 98 | |||
| DQ890037.1 | - | 172 | 98 | |||
| DQ890038.1 | - | 125 | 98 | |||
| DQ890039.1 | - | 165 | 98 | |||
| DQ890040.1 | - | 165 | 98 | |||
| DQ890041.1 | - | 165 | 98 | |||
| DQ890042.1 | - | 147 | 98 | |||
| DQ890043.1 | - | 165 | 98 | |||
| DQ890044.1 | - | 165 | 98 | |||
| DQ890045.1 | - | 165 | 98 | |||
| DQ890046.1 | - | 165 | 98 | |||
| DQ890047.1 | - | 165 | 98 | |||
| DQ890048.1 | - | 165 | 98 | |||
| DQ890049.1 | - | 165 | 98 | |||
| DQ890050.1 | - | 165 | 98 | |||
| DQ890051.1 | - | 165 | 98 | |||
| DQ890052.1 | - | 165 | 98 | |||
| DQ890053.1 | - | 165 | 98 | |||
| DQ890054.1 | - | 165 | 98 | |||
| DQ890055.1 | - | 165 | 98 | |||
| DQ890056.1 | - | 165 | 98 | |||
| DQ890057.1 | - | 165 | 98 | |||
| DQ890058.1 | - | 165 | 98 | |||
| DQ890059.1 | - | 138 | 98 | |||
| DQ890060.1 | - | 165 | 98 | |||
| DQ890061.1 | - | 165 | 98 | |||
| DQ890062.1 | - | 165 | 98 | |||
| DQ890063.1 | - | 165 | 98 | |||
| L26029.1 | - | 172 | 98 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

IL10
Genomic Map