Assay Details
| Gene Symbol: CDC20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261117.1 | NP_001248046.1 | 1173 | 109 | ||
| XM_028837855.1 | XP_028693688.1 | 2626 | 109 | |||
| GenBank mRNA | JU320900.1 | - | 1145 | 109 | ||
| JV045155.1 | - | 1173 | 109 | |||
| JV666994.1 | - | 1192 | 109 | |||
| JV728112.1 | - | 109 | ||||
More Information
Set Membership: |
|

CDC20
Genomic Map