Assay Details
| Gene Symbol: ZFAND2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260713.1 | NP_001247642.1 | 658 | 57 | ||
| XM_015110987.2 | XP_014966473.2 | 699 | 57 | |||
| XM_015110988.2 | XP_014966474.2 | 699 | 57 | |||
| XM_028830474.1 | XP_028686307.1 | 659 | 57 | |||
| XM_028830475.1 | XP_028686308.1 | 659 | 57 | |||
| XM_028830476.1 | XP_028686309.1 | 559 | 57 | |||
| GenBank mRNA | JU322995.1 | - | 666 | 57 | ||
| JU473744.1 | - | 701 | 57 | |||
| JV045520.1 | - | 658 | 57 | |||
| JV638482.1 | - | 712 | 57 | |||
More Information
Set Membership: |
|

ZFAND2B
Genomic Map