Assay Details
| Gene Symbol: HIVEP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001085309.4 | XP_001085309.2 | 6242 | 65 | ||
| XM_015136908.2 | XP_014992394.2 | 5986 | 65 | |||
| XM_028835187.1 | XP_028691020.1 | 5862 | 65 | |||
| XM_028835195.1 | XP_028691028.1 | 6634 | 65 | |||
| GenBank mRNA | JU476642.1 | - | 5937 | 65 | ||
More Information
Set Membership: |
|

HIVEP3
Genomic Map