Assay Details
| Gene Symbol: ACBD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001115247.4 | XP_001115247.1 | 1020 | 56 | ||
| XM_015119904.2 | XP_014975390.1 | 929 | 56 | |||
| XM_015119905.2 | XP_014975391.1 | 924 | 56 | |||
| XM_015119906.2 | XP_014975392.1 | 913 | 56 | |||
| XM_028836610.1 | XP_028692443.1 | 415 | 56 | |||
| XM_028836611.1 | XP_028692444.1 | 398 | 56 | |||
More Information
Set Membership: |
|

ACBD4
Genomic Map