Assay Details
| Gene Symbol: DIAPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001087743.4 | XP_001087743.1 | 1427 | 99 | ||
| XM_001087859.4 | XP_001087859.1 | 1405 | 99 | |||
| XM_001087983.4 | XP_001087983.1 | 1406 | 99 | |||
| XM_015127777.2 | XP_014983263.1 | 1427 | 99 | |||
| XM_028841927.1 | XP_028697760.1 | 1393 | 99 | |||
More Information
Set Membership: |
|

DIAPH2
Genomic Map