Assay Details
| Gene Symbol: BLZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125013.2 | XP_014980499.1 | 1170 | 97 | ||
| XM_015125021.2 | XP_014980507.1 | 538 | 97 | |||
| XM_028850182.1 | XP_028706015.1 | 598 | 97 | |||
| GenBank mRNA | JU336850.1 | - | 769 | 97 | ||
| JU477594.1 | - | 866 | 97 | |||
More Information
Set Membership: |
|

BLZF1
Genomic Map