Assay Details
| Gene Symbol: XBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190861.2 | NP_001177790.1 | 611 | 66 | ||
| NM_001271739.1 | NP_001258668.1 | 585 | 66 | |||
| GenBank mRNA | CN645482.1 | - | 282 | 66 | ||
| CO647703.1 | - | 615 | 66 | |||
| JU336259.1 | - | 595 | 66 | |||
| JU336260.1 | - | 621 | 66 | |||
| JV048174.1 | - | 618 | 66 | |||
| JV627892.1 | - | 643 | 66 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

XBP1
Genomic Map