Assay Details
| Gene Symbol: LOC114677617 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028847619.1 | XP_028703452.1 | 190 | 83 | ||
| Gene Symbol: STK38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260590.1 | NP_001247519.1 | 1102 | 83 | ||
| GenBank mRNA | JU474200.1 | - | 1102 | 83 | ||
More Information
Set Membership: |
|

LOC114677617
Genomic Map