Assay Details
| Gene Symbol: AGGF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261037.1 | NP_001247966.1 | 1981 | 80 | ||
| GenBank mRNA | JU335228.1 | - | 1972 | 80 | ||
| JU476192.1 | - | 1981 | 80 | |||
| JV047500.1 | - | 1976 | 80 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

AGGF1
Genomic Map