Assay Details
| Gene Symbol: MBIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015143578.2 | XP_014999064.1 | 621 | 113 | ||
| XM_015143579.2 | XP_014999065.1 | 621 | 113 | |||
| XM_015143581.2 | XP_014999067.1 | 621 | 113 | |||
| XM_015143582.2 | XP_014999068.1 | 621 | 113 | |||
| XM_028851501.1 | XP_028707334.1 | 621 | 113 | |||
| XM_028851502.1 | XP_028707335.1 | 1024 | 113 | |||
| XM_028851503.1 | XP_028707336.1 | 621 | 113 | |||
| GenBank mRNA | JV045637.1 | - | 524 | 113 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MBIP
Genomic Map