Assay Details
| Gene Symbol: CST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032924.2 | NP_001028096.1 | 295 | 93 | ||
| GenBank mRNA | AY885223.1 | - | 300 | 93 | ||
| JV534274.1 | - | 228 | 93 | |||
| U51912.1 | - | 240 | 93 | |||
| Gene Symbol: RPSA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JV616428.1 | - | 918 | 93 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CST3
Genomic Map