Assay Details
| Gene Symbol: ATP5F1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195494.1 | NP_001182423.1 | 806 | 114 | ||
| GenBank mRNA | DR772444.1 | - | 453 | 114 | ||
| JU487516.1 | - | 684 | 114 | |||
| JV470111.1 | - | 766 | 114 | |||
| JV583257.1 | - | 767 | 114 | |||
| Gene Symbol: LOC705465 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU326567.1 | - | 774 | 114 | ||
More Information
Set Membership: |
|

ATP5F1B
Genomic Map