Assay Details
| Gene Symbol: NARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001094498.4 | XP_001094498.2 | 1340 | 95 | ||
| XM_015115337.2 | XP_014970823.1 | 1340 | 95 | |||
| XM_028833867.1 | XP_028689700.1 | 1335 | 95 | |||
| GenBank mRNA | JU323283.1 | - | 1277 | 95 | ||
| JU471336.1 | - | 1286 | 95 | |||
| JV044886.1 | - | 1246 | 95 | |||
| JV703755.1 | - | 1291 | 95 | |||
More Information
Set Membership: |
|

NARS2
Genomic Map