Assay Details
| Gene Symbol: CTSO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015139410.1 | XP_014994896.1 | 622 | 113 | ||
| GenBank mRNA | JV046275.1 | - | 616 | 113 | ||
| JV643398.1 | - | 519 | 113 | |||
| JV707867.1 | - | 528 | 113 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CTSO
Genomic Map