Assay Details
| Gene Symbol: TRIM44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260523.1 | NP_001247452.1 | 1312 | 61 | ||
| GenBank mRNA | JU476111.1 | - | 1306 | 61 | ||
| JV044348.1 | - | 1312 | 61 | |||
More Information
Set Membership: |
|

TRIM44
Genomic Map