Assay Details
Gene Symbol: CLU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195474.1 | NP_001182403.1 | 1235 | 93 | ||
XM_015144999.2 | XP_015000485.1 | 1514 | 93 | |||
XM_028852103.1 | XP_028707936.1 | 1566 | 93 | |||
XM_028852104.1 | XP_028707937.1 | 1601 | 93 | |||
XM_028852105.1 | XP_028707938.1 | 1484 | 93 | |||
GenBank mRNA | CN647010.1 | - | 220 | 93 | ||
JV532915.1 | - | 1344 | 93 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
