Assay Details
| Gene Symbol: NOP56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149408.2 | XP_015004894.1 | 1391 | 59 | ||
| XM_015149410.2 | XP_015004896.1 | 1391 | 59 | |||
| GenBank mRNA | JU321410.1 | - | 950 | 59 | ||
| JV567508.1 | - | 957 | 59 | |||
| JV703564.1 | - | 959 | 59 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NOP56
Genomic Map