Assay Details
| Gene Symbol: LAMTOR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257535.1 | NP_001244464.1 | 515 | 75 | ||
| GenBank mRNA | JU320556.1 | - | 515 | 75 | ||
| JU471004.1 | - | 479 | 75 | |||
| JV044392.1 | - | 495 | 75 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LAMTOR3
Genomic Map