Assay Details
| Gene Symbol: NR4A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002800032.3 | XP_002800078.2 | 1982 | 102 | ||
| XM_028835023.1 | XP_028690856.1 | 2162 | 102 | |||
More Information
Set Membership: |
|

NR4A3
Genomic Map