Assay Details
| Gene Symbol: SYDE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123165.2 | XP_014978651.1 | 1482 | 63 | ||
| XM_028840067.1 | XP_028695900.1 | 1281 | 63 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SYDE1
Genomic Map