Assay Details
| Gene Symbol: OAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083949.1 | NP_001077418.1 | 659 | 65 | ||
| GenBank mRNA | EF467665.1 | - | 665 | 65 | ||
| EF467666.1 | - | 659 | 65 | |||
| JV638063.1 | - | 736 | 65 | |||
| MH188460.1 | - | 665 | 65 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

OAS1
Genomic Map