Assay Details
| Gene Symbol: PRKAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149729.2 | XP_015005215.1 | 544 | 105 | ||
| XM_015149739.2 | XP_015005225.1 | 1309 | 105 | |||
| GenBank mRNA | JU327297.1 | - | 400 | 105 | ||
| JU476324.1 | - | 563 | 105 | |||
| JV045430.1 | - | 452 | 105 | |||
| JV637828.1 | - | 556 | 105 | |||
More Information
Set Membership: |
|

PRKAB2
Genomic Map