Assay Details
| Gene Symbol: CDK9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266230.1 | NP_001253159.1 | 656 | 103 | ||
| GenBank mRNA | JU332460.1 | - | 660 | 103 | ||
| JU473926.1 | - | 656 | 103 | |||
| JV637371.1 | - | 944 | 103 | |||
More Information
Set Membership: |
|

CDK9
Genomic Map