Assay Details
| Gene Symbol: SIRT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015135578.2 | XP_014991064.2 | 704 | 83 | ||
| XM_015135579.2 | XP_014991065.2 | 760 | 83 | |||
| XM_015135582.2 | XP_014991068.2 | 672 | 83 | |||
| XM_028847824.1 | XP_028703657.1 | 882 | 83 | |||
| XM_028847825.1 | XP_028703658.1 | 964 | 83 | |||
| XM_028847826.1 | XP_028703659.1 | 818 | 83 | |||
| XM_028847827.1 | XP_028703660.1 | 828 | 83 | |||
| XM_028847828.1 | XP_028703661.1 | 799 | 83 | |||
| XM_028847829.1 | XP_028703662.1 | 779 | 83 | |||
| XM_028847830.1 | XP_028703663.1 | 877 | 83 | |||
| XM_028847831.1 | XP_028703664.1 | 765 | 83 | |||
| XM_028847832.1 | XP_028703665.1 | 704 | 83 | |||
| XM_028847833.1 | XP_028703666.1 | 680 | 83 | |||
| GenBank mRNA | JU483468.1 | - | 611 | 83 | ||
More Information
Set Membership: |
|

SIRT5
Genomic Map