Assay Details
| Gene Symbol: DCXR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001112983.4 | XP_001112983.2 | 515 | 54 | ||
| XM_028837015.1 | XP_028692848.1 | 517 | 54 | |||
| GenBank mRNA | JU323147.1 | - | 457 | 54 | ||
| JV716359.1 | - | 448 | 54 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DCXR
Genomic Map