Assay Details
| Gene Symbol: BRD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194118.2 | NP_001181047.1 | 2078 | 90 | ||
| XM_015116407.2 | XP_014971893.1 | 2532 | 90 | |||
| XM_028834368.1 | XP_028690201.1 | 2188 | 90 | |||
| XM_028834369.1 | XP_028690202.1 | 2072 | 90 | |||
| GenBank mRNA | JU477296.1 | - | 2119 | 90 | ||
| JV048303.1 | - | 2126 | 90 | |||
| JV636386.1 | - | 2119 | 90 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

BRD3
Genomic Map