Assay Details
| Gene Symbol: NMNAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122174.2 | XP_014977660.1 | 758 | 51 | ||
| GenBank mRNA | JU324450.1 | - | 320 | 51 | ||
| JV667252.1 | - | 269 | 51 | |||
More Information
Set Membership: |
|

NMNAT2
Genomic Map