Assay Details
| Gene Symbol: CBX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150376.2 | XP_015005862.1 | 248 | 62 | ||
| XM_015150377.2 | XP_015005863.1 | 248 | 62 | |||
| GenBank mRNA | JV576641.1 | - | 176 | 62 | ||
More Information
Set Membership: |
|

CBX6
Genomic Map