Assay Details
| Gene Symbol: DUSP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015114009.2 | XP_014969495.1 | 1627 | 65 | ||
| XM_015114018.2 | XP_014969504.1 | 1426 | 65 | |||
| GenBank mRNA | JU332758.1 | - | 872 | 65 | ||
| JU474172.1 | - | 871 | 65 | |||
| JU479548.1 | - | 884 | 65 | |||
More Information
Set Membership: |
|

DUSP12
Genomic Map