Assay Details
| Gene Symbol: DIO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001324507.3 | NP_001311436.2 | 270 | 84 | ||
| GenBank mRNA | JU332319.1 | - | 270 | 84 | ||
| JV573354.1 | - | 64 | 84 | |||
More Information
Set Membership: |
|

DIO2
Genomic Map