Assay Details
| Gene Symbol: ONECUT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122065.2 | XP_014977551.2 | 1598 | 59 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ONECUT2| Gene Symbol: ONECUT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122065.2 | XP_014977551.2 | 1598 | 59 | ||
Set Membership: |
|