Assay Details
| Gene Symbol: CORO2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015117227.2 | XP_014972713.2 | 1080 | 66 | ||
| GenBank mRNA | JU492857.1 | - | 866 | 66 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CORO2A| Gene Symbol: CORO2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015117227.2 | XP_014972713.2 | 1080 | 66 | ||
| GenBank mRNA | JU492857.1 | - | 866 | 66 | ||
Set Membership: |
|