Assay Details
| Gene Symbol: ZC3H10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001114150.4 | XP_001114150.3 | 220 | 74 | ||
| XM_015151968.2 | XP_015007454.1 | 452 | 74 | |||
| GenBank mRNA | JU320426.1 | - | 56 | 74 | ||
| JU470900.1 | - | 118 | 74 | |||
| JV631344.1 | - | 101 | 74 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001114150.4, XM_015151968.2 transcripts.
Set Membership: |
|

ZC3H10
Genomic Map