Assay Details
| Gene Symbol: FGGY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015140756.2 | XP_014996242.2 | 1120 | 99 | ||
| XM_015140764.2 | XP_014996250.2 | 1034 | 99 | |||
| XM_015140794.2 | XP_014996280.2 | 1089 | 99 | |||
| XM_015140820.2 | XP_014996306.2 | 467 | 99 | |||
| XM_028833163.1 | XP_028688996.1 | 1628 | 99 | |||
| XM_028833165.1 | XP_028688998.1 | 1442 | 99 | |||
| XM_028833175.1 | XP_028689008.1 | 1670 | 99 | |||
| XM_028833179.1 | XP_028689012.1 | 1456 | 99 | |||
| XM_028833190.1 | XP_028689023.1 | 1034 | 99 | |||
| XM_028833196.1 | XP_028689029.1 | 812 | 99 | |||
| XM_028833199.1 | XP_028689032.1 | 866 | 99 | |||
| GenBank mRNA | JU324189.1 | - | 908 | 99 | ||
| JV064982.1 | - | 1017 | 99 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028833199.1 transcripts.
Set Membership: |
|

FGGY
Genomic Map