Assay Details
| Gene Symbol: HMGCS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001114146.4 | XP_001114146.2 | 823 | 62 | ||
| XM_015149389.2 | XP_015004875.2 | 823 | 62 | |||
| GenBank mRNA | JV635673.1 | - | 648 | 62 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HMGCS2
Genomic Map