Assay Details
| Gene Symbol: DAXX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136105.2 | XP_014991591.2 | 2190 | 129 | ||
| XM_015136106.2 | XP_014991592.2 | 2163 | 129 | |||
| XM_015136107.2 | XP_014991593.2 | 2096 | 129 | |||
| XM_028847679.1 | XP_028703512.1 | 1866 | 129 | |||
| GenBank mRNA | JU339569.1 | - | 588 | 129 | ||
| JU474489.1 | - | 2109 | 129 | |||
| JV046306.1 | - | 2092 | 129 | |||
| JV635985.1 | - | 2106 | 129 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DAXX
Genomic Map