Assay Details
| Gene Symbol: GALNT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194577.2 | NP_001181506.1 | 542 | 87 | ||
| XM_028834389.1 | XP_028690222.1 | 542 | 87 | |||
| GenBank mRNA | JU493509.1 | - | 317 | 87 | ||
| JV661526.1 | - | 303 | 87 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GALNT12
Genomic Map