Assay Details
| Gene Symbol: NRAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148825.2 | XP_015004311.1 | 1178 | 93 | ||
| GenBank mRNA | JU334698.1 | - | 573 | 93 | ||
| JU470736.1 | - | 552 | 93 | |||
| JV043759.1 | - | 557 | 93 | |||
More Information
Set Membership: |
|

NRAS
Genomic Map