Assay Details
| Gene Symbol: COL21A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136589.2 | XP_014992075.2 | 2522 | 75 | ||
| XM_015136590.2 | XP_014992076.2 | 2352 | 75 | |||
| XM_028847464.1 | XP_028703297.1 | 1805 | 75 | |||
| XM_028847465.1 | XP_028703298.1 | 781 | 75 | |||
| XM_028847466.1 | XP_028703299.1 | 317 | 75 | |||
More Information
Set Membership: |
|

COL21A1
Genomic Map