Assay Details
| Gene Symbol: VPS52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136102.2 | XP_014991588.1 | 1523 | 77 | ||
| XM_028847680.1 | XP_028703513.1 | 1523 | 77 | |||
| GenBank mRNA | JU321816.1 | - | 1487 | 77 | ||
| JU472314.1 | - | 1484 | 77 | |||
| JV043922.1 | - | 1488 | 77 | |||
| JV529184.1 | - | 1490 | 77 | |||
More Information
Set Membership: |
|

VPS52
Genomic Map