Assay Details
| Gene Symbol: DGCR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194750.1 | NP_001181679.1 | 2002 | 62 | ||
| XM_002798275.3 | XP_002798321.1 | 2039 | 62 | |||
| XM_015149717.2 | XP_015005203.1 | 4509 | 62 | |||
| XM_015149719.2 | XP_015005205.1 | 2039 | 62 | |||
| GenBank mRNA | JU327586.1 | - | 1166 | 62 | ||
| JU472935.1 | - | 1978 | 62 | |||
| JV047434.1 | - | 1980 | 62 | |||
| JV472045.1 | - | 1320 | 62 | |||
| JV667419.1 | - | 1607 | 62 | |||
More Information
Set Membership: |
|

DGCR8
Genomic Map