Assay Details
| Gene Symbol: HS6ST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015109806.2 | XP_014965292.2 | 830 | 101 | ||
| GenBank mRNA | JU328613.1 | - | 523 | 101 | ||
| JU477887.1 | - | 552 | 101 | |||
| JU477888.1 | - | 552 | 101 | |||
| JV565550.1 | - | 556 | 101 | |||
More Information
Set Membership: |
|

HS6ST1
Genomic Map