Assay Details
| Gene Symbol: ELK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194388.2 | NP_001181317.1 | 1118 | 57 | ||
| XM_015152478.2 | XP_015007964.1 | 1333 | 57 | |||
| XM_028828962.1 | XP_028684795.1 | 1025 | 57 | |||
| GenBank mRNA | JU471176.1 | - | 1266 | 57 | ||
| JV046394.1 | - | 1155 | 57 | |||
| JV636013.1 | - | 1184 | 57 | |||
More Information
Set Membership: |
|

ELK3
Genomic Map