Assay Details
| Gene Symbol: CHST11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015152630.2 | XP_015008116.1 | 623 | 90 | ||
| GenBank mRNA | JU328116.1 | - | 73 | 90 | ||
| JU475778.1 | - | 614 | 90 | |||
More Information
Set Membership: |
|

CHST11
Genomic Map