Assay Details
| Gene Symbol: MFAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266342.1 | NP_001253271.1 | 395 | 83 | ||
| GenBank mRNA | JU334559.1 | - | 474 | 83 | ||
| JU473764.1 | - | 468 | 83 | |||
| JV045164.1 | - | 395 | 83 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MFAP1
Genomic Map