Assay Details
| Gene Symbol: IL17RD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001100582.4 | XP_001100582.1 | 292 | 68 | ||
| XM_028844287.1 | XP_028700120.1 | 212 | 68 | |||
| GenBank mRNA | JV654464.1 | - | 107 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028844287.1 transcripts.
Set Membership: |
|

IL17RD
Genomic Map